Please login to your online account to display your discounted pricing

Wheaton™ Polypropylene Caps for Wheaton Dropping Bottles - Purple


Manufacturer: wheaton™  W242517A

 View more versions of this product

Catalog No. 03-440-07

  • / Case of 1000 Each
 

Description & Specifications

Specifications

Neck Finish 15-415
For Use With (Equipment) Plastic dropping bottle
Certifications/Compliance ISO 9001
Color Purple
Material Polypropylene
No. per Case 1000 Cs.