Please login to your online account to display your discounted pricing

Wheaton™ Polypropylene Caps for Wheaton Dropping Bottles - Gray


Manufacturer: wheaton™  W242535A

 View more versions of this product

Catalog No. 03-439-87

  • / Case of 1000 Each
 

Description & Specifications

Specifications

Neck Finish 13-425
Material Polypropylene
Certifications/Compliance ISO 9001
Color Gray
No. per Case 1000/Cs.
For Use With Plastic dropping bottle