Please login to your online account to display your discounted pricing

Wheaton™ Polypropylene Caps for Wheaton Dropping Bottles - Gray


Manufacturer: wheaton™  W242525A

 View more versions of this product

Catalog No. 03-439-98

  • / Case of 1000 Each
 

Description & Specifications

Specifications

Certifications/Compliance ISO 9001
Color Gray
Material Polypropylene
Neck Finish 20-410
For Use With Plastic dropping bottle
No. per Case 1000/Cs.