missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Wheaton™ Polypropylene Caps for Wheaton Dropping Bottles - Blue

Screw cap made of radiation grade polypropylene

$36.50 - $659.25

Specifications

Certifications/Compliance ISO 9001
Color Blue
Material Polypropylene
For Use With (Equipment) Plastic dropping bottle
View More Specs

Products 5
Catalog Number Mfr. No. Neck Finish No. per Case Price Quantity  
Catalog Number Mfr. No. Neck Finish No. per Case Price Quantity  
03-439-99
View Documents Promotion Details
DWK Life Sciences
242524A
20-410 1000/Cs.
Case of 1000 Each for $659.25
 
06-450-152
View Documents Promotion Details
DWK Life Sciences
242504
8-425 100/Cs.
Case of 100 Each for $36.50
 
03-440-20
View Documents Promotion Details
DWK Life Sciences
242504A
8-425 1000/Cs.
Case of 1000 Each for $397.12
 
03-440-10
View Documents Promotion Details
DWK Life Sciences
242514A
15-415 1000/Cs.
Case of 1000 Each for $634.50
 
03-439-88
View Documents Promotion Details
DWK Life Sciences
242534A
13-425 1000/Cs.
Case of 1000 Each for $539.92
 
Description

Description

  • Packaged in case of 100 or 1000
Specifications

Specifications

ISO 9001
Blue
Polypropylene
Plastic dropping bottle
SDS
Product Certifications