DWK Life Sciences Wheaton™ Polypropylene Caps for S/T Bottles
For use with Wheaton Glass Diagnostic Bottles
Manufacturer: DWK Life Sciences 24071601
Description
- Unlined
- Open- and closed-top caps
- 20-400 screw thread
- Autoclavable
Specifications
ISO 9001 | |
Screw neck diagnostic bottle | |
Polypropylene | |
20-400 |
Open Top Cap | |
Black | |
300 Cs. |