DWK Life Sciences Wheaton™ Polyacetal Clamps
Manufacturer:  DWK Life Sciences W297749
 View more versions of this product
Catalog No. 05-884D
Description
Description
  • Molded from polyoxymethylene (POM), an acetal resin
  • Will not scratch glass and can withstand temperatures up to 150°C
  • Resistant to bases and weak acids
  • Not resistant to halogen gases and concentrated acids
Specifications
Specifications
Blue
Kuderna Danish Evaporative Concentrator
POM Acetal
Standard Taper Joint Clamp