DWK Life Sciences Wheaton™ Polyacetal Clamps
Color-coded clamps hold standard taper joints securely under moderate pressure
Supplier: DWK Life Sciences 297746
Description
- Molded from polyoxymethylene (POM), an acetal resin
- Will not scratch glass and can withstand temperatures up to 150°C
- Resistant to bases and weak acids
- Not resistant to halogen gases and concentrated acids
Specifications
Standard Taper Clamp | |
POM Acetal | |
Kuderna Danish Evaporative Concentrator | |
Yellow |