missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Wheaton™ Plastic Nongraduated Media Bottles

Sterile and versatile

$348.06 - $348.06

Specifications

For Use With (Application) For distilled Water, analytical standards and reagents when fitted with a Polyethylene lined screw cap
Closure Material Polypropylene
Closure Size 38-430
Color Clear
Material PET
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (English) Height (English) Quantity Price Quantity  
Catalog Number Mfr. No. Diameter (English) Height (English) Quantity Price Quantity  
06-451-298 Wheaton™
219975
2.08 in. 3.89 in. 24 Cs. Case of 48 Each for $348.06
Specifications

Description

Ideal for mixing and storing culture media, or for use with distilled water, analytical standards and reagents when fitted with a polyethylene-lined screw cap.

 

  • Large opening, special lip and sloping shoulders for easy pouring, cleaning
  • With polyethylene-lined caps
  • Supplied with caps pre-attached and shrink-wrapped in convenient trays

 

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

For distilled Water, analytical standards and reagents when fitted with a Polyethylene lined screw cap
38-430
PET
Square
No
Polypropylene
Clear
33-430
Sterile