DWK Life Sciences Wheaton™ Safety-Coated Narrow Mouth Bottles with Aluminum-Foil-Lined Black Phenolic Caps
Retain contents and most glass fragments for safer and easier disposal
$31.77 - $31.77
Specifications
Certifications/Compliance USP Type III
Closure Included Yes
Closure Liner Aluminum Foil
Closure Material Phenolic
Coating Plastisol
View More Specs
Products 1
Catalog Number Mfr. No. Capacity Color Height (Metric) Height (English) Price Quantity  
Catalog Number Mfr. No. Capacity Color Height (Metric) Height (English) Price Quantity  
03-439-50A
View Documents
DWK Life Sciences
220738
80 oz., 2300 mL Clear 295mm 11.6 in. Each for $31.77
Case of 6 Each for $190.62
Description
Description
Retain contents and most glass fragments for safer and easier disposal
  • PVC coated
  • Coating is inert and highly resistant to strong acids, alkalis, and most solvents
  • Coating reduces risk of personal injury or contamination
  • Black, foil-lined caps included
  • Screw cap size: 38mm-430
  • Not autoclavable
Compliance
Helps meet OSHA requirements for safe handling and disposal of toxic or noxious substances.
Specifications
Specifications
USP Type III
Aluminum Foil
Plastisol
Black
Yes
Phenolic
Screw
Plastisol Coated Soda-Lime Glass