DWK Life Sciences Wheaton™ Open Top Screw Caps with Silicone Liner
Open top screw caps have a bonded PTFE faced silicone liner that provides access with a syringe.
Supplier: DWK Life Sciences W240848
Description
- Can withstand autoclaving / sterilization
Specifications
Open Top Screw Cap | |
Glass bottle or vial | |
Phenolic Resin | |
13-425 |
PTFE/Silicone | |
White | |
250 Cs. | |
ISO 9001 |