Please login to your online account to display your discounted pricing

Wheaton™ Open Top Screw Caps with Silicone Liner


Manufacturer: wheaton™  W240849

 View more versions of this product

Catalog No. 03-410-043

  • / Case of 1000 Each
 

Description & Specifications

Specifications

Certifications/Compliance ISO 9001
Closure Liner PTFE/Silicone
Color Black
No. per Case 1000 Cs.
For Use With (Equipment) Glass bottle or vial
Neck Finish 13-425