DWK Life Sciences Wheaton™ Omnispense™ Plus Peristaltic Pump Accessory Pump Heads
For use with Wheaton Omnispense Plus dispensing pump
Manufacturer:  DWK Life Sciences W375025
 View more versions of this product
Catalog No. 13-687-445
Description
Description
  • Stackable pump heads (13-687-444 and -442) allow user to add up to two additional pumpheads, for a total of three pump heads per dispensing pump
  • Cassette pump head ideal for multichannel applications
  • Removable cassettes can be preloaded with tubing elements and changed without stopping the drive
†For tubing with bore sizes from 0.13 to 2.79mm.
Specifications
Specifications
Cassette Pump Head
Omnispense™ plus peristaltic pump
4.0
5
Documents