DWK Life Sciences Wheaton™ Microlink Caps
Open top caps are ideal for storing and containing volatile solvents, sensitive compounds and corrosive chemicals
Manufacturer:  DWK Life Sciences 240106
 View more versions of this product
Catalog No. 06-450-10
Description
Description
Open top caps are ideal for storing and containing volatile solvents, sensitive compounds and corrosive chemicals
  • Polypropylene/silicone/PTFE liner is ultrasonically welded to each cap to help eliminate possibility of toxicity
  • Autoclavable
Specifications
Specifications
Autoclavable
Polypropylene/Silicone
Black
50 Cs.
ISO 9001
Bottle
Polypropylene
33-430