DWK Life Sciences Wheaton™ Microlink Caps
Open top caps are ideal for storing and containing volatile solvents, sensitive compounds and corrosive chemicals
$114.15 - $397.29
Specifications
For Use With (Equipment) Bottle
Material Polypropylene
Certifications/Compliance ISO 9001
View More Specs
Products 3
Catalog Number Mfr. No. Neck Finish Color No. per Case Price Quantity  
Catalog Number Mfr. No. Neck Finish Color No. per Case Price Quantity  
06-450-10
View Documents Promotion Details
DWK Life Sciences
240106
33-430 Black 50 Cs.
Pack of 50 Each for $114.15
06-450-12
View Documents Promotion Details
DWK Life Sciences
240112
33-430 Black 50 Cs.
Case of 50 Each for $294.88
06-450-100
View Documents Promotion Details
DWK Life Sciences
24025601
38-430 White 24 Cs.
Case of 24 Each for $397.29
Description
Description
Open top caps are ideal for storing and containing volatile solvents, sensitive compounds and corrosive chemicals
  • Polypropylene/silicone/PTFE liner is ultrasonically welded to each cap to help eliminate possibility of toxicity
  • Autoclavable
Specifications
Specifications
Bottle
Polypropylene
ISO 9001
SDS
missing translation for 'documents'
missing translation for 'documents'