Wheaton™ Magnetic Stirrers
Manufacturer:  Wheaton™ WW900702A
 View more versions of this product
Catalog No. 02-912-639
Specifications
Description
Stirrer, Magnetic; Wheaton; BioStir; For general mixing; 150-1200rpm; Programmable; Remote controlled; Alarm capable; Plug Style: 120VAC North America; 1 bottle positions; 1/Cs.
  • Permanent fluid path visibility
  • Integrated calibration system
  • Choice of adjustment button with each unit
  • Long lasting performance stability
  • Autoclavable at 121°C / 250°F fully assembled
  • Two year warranty
Description & Specifications
Specifications
120VAC North America
150 to 1200RPM
Magnetic Stirrers