missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Wheaton™ Magna-Flex™ Spinner Flasks
Produce high-yield cultures by maintaining even circulation while keeping microcarriers in suspension
Manufacturer: DWK Life Sciences 356839
Description
- Borosilicate glass flasks
- Two large side arms with screw-cap closures
- Flex-type, bulb-shaped glass impeller rotates from a fixed position above the liquid level around an indentation in the base of the flask
- Graduated
- Removable stainless-steel pin immobilizes the impeller during handling or decanting to prevent damage to cells or microcarriers
- Autoclavable flasks and impellers for selected sizes can also be ordered separately
- U.S. Patent No. 3,854,704
Specifications
4.5 cm | |
Borosilicate Glass | |
Autoclavable |
6 L | |
Flask with impeller | |
Yes |