DWK Life Sciences Wheaton™ M-T Vial File™ and Accessories
For compact and organized sample storage
Manufacturer: DWK Life Sciences 228778*
Description
For compact and organized sample storage
- Stores vials in clear polystyrene case
- Foam inserts and partitions provide protection
- With alphanumerical indexing card for identification
- Index cards and partitions also available separately
Specifications
Vial storage and transport | |
Clear polystyrene hinged case, partitions, foam inserts and alphanumerical indexing card | |
M-T Vial File | |
Polystyrene |
60 x 2mL vials | |
60 | |
6 Case |