Please login to your online account to display your discounted pricing
Wheaton™ M-T Vial File™ and Accessories
For compact and organized sample storage
Manufacturer: wheaton™  WW228788
 View more versions of this product
Catalog No. 02-912-825
  • / Case of 6 Each
Description & Specifications
Specifications
For Use With (Equipment) 20mL vials M-T vial file
For Use With (Application) Vial storage and transport
Quantity 6 Case
Product Type M-T Vial File Partition
  • Stores vials in clear polystyrene case
  • Foam inserts and partitions provide protection
  • With alphanumerical indexing card for identification
  • Index cards and partitions also available separately