DWK Life Sciences Wheaton™ M-T Vial File™ and Accessories
For compact and organized sample storage
Supplier: DWK Life Sciences W228786
Description
For compact and organized sample storage
- Stores vials in clear polystyrene case
- Foam inserts and partitions provide protection
- With alphanumerical indexing card for identification
- Index cards and partitions also available separately
Specifications
Storage and Transportation of Sample Vials | |
W228790 Hinged Case | |
Partition | |
6 Case |