Wheaton™ M-T Vial File™ and Accessories
For compact and organized sample storage
Manufacturer:  Wheaton™ WW228789
 View more versions of this product
Catalog No. 02-912-823
Specifications
Description
Card, Index; Wheaton; For 20mL M-T vial file
  • Stores vials in clear polystyrene case
  • Foam inserts and partitions provide protection
  • With alphanumerical indexing card for identification
  • Index cards and partitions also available separately
Description & Specifications
Specifications
Vial storage and transport
20mL M-T vial file
Alphanumerical Index Card
35 Case