Please login to your online account to display your discounted pricing

Wheaton™ Lyophilized Stopper

$542.58 - $821.00

Specifications

Autoclavable Yes
Diameter (Metric) Inside 1.3cm
Color Gray
Material Bromobutyl
For Use With (Application) Lyophilization
View More Specs
Products
Catalog Number Mfr. No. Product Type Price Quantity    

02-913-552

 
wheaton™
WW224100402
Straight Plug Stopper Case of 1000 Each for $542.58

02-913-555

 
wheaton™
WW224100406
2-Leg Lyophilization Stopper Case of 1000 Each for $821.00

02-913-556

 
wheaton™
WW224100407
3-Leg Lyophilization Stopper Case of 1000 Each for $748.21

02-913-557

 
wheaton™
WW224100408
Igloo Lyophilization Stopper Case of 1000 Each for $821.00
Description & Specifications

Specifications

Autoclavable Yes
Diameter (Metric) Inside 1.3cm
Color Gray
Material Bromobutyl
For Use With (Application) Lyophilization
Hardness 46 Shore A

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.