Please login to your online account to display your discounted pricing

Wheaton™ Lyophilized Stopper


Manufacturer: wheaton™  WW224100407

 View more versions of this product

Catalog No. 02-913-556

  • / Case of 1000 Each
 

Description & Specifications

Specifications

Autoclavable Yes
Diameter (English) Outside 0.78 in.
Diameter (Metric) Inside 1.3cm
Diameter (Metric) Outside 2cm
For Use With (Application) Lyophilization
Hardness 46 Durometer
Color Gray
Material Bromobutyl
Product Type 3-Leg Lyophilization Stopper