Wheaton™ Low-Density Extended Controlled Dropper Tips (15mm)
Manufacturer:  Wheaton™ W242421A
 View more versions of this product
Catalog No. 03-440-46
Specifications
Description
Tips, Dropper; Wheaton; Low-density; Polyethylene; Extended; Controlled; Size: 15mm; Color: Pink; 1000/Cs.
  • Extended controlled dropper tip dispenses one drop at a time
  • Average drop size is 44μL ±6μL
  • Case of 1000
Description & Specifications
Specifications
ISO 9001
41μL
1000 Cs.
Plastic dropping bottle
Pink
Polyethylene
Dropper Tips