DWK Life Sciences Wheaton™ Low-Density Extended Controlled Dropper Tips (15mm)
Manufacturer:  DWK Life Sciences W242404A
 View more versions of this product
Catalog No. 03-440-63B
Description
Description
  • Extended controlled dropper tip dispenses one drop at a time
  • Average drop size is 44μL ±6μL
  • Case of 1000
Specifications
Specifications
ISO 9001
41μL
1000 Cs.
Plastic dropping bottle
Blue
Polyethylene
Dropper Tips
Documents