Wheaton™ Low-Density Extended Controlled Dropper Tips (15mm)
Manufacturer:  Wheaton™ W242408A
 View more versions of this product
Catalog No. 03-440-60
Specifications
Description
Caps, Extended Controlled Dropper Tip; Wheaton; LDPE; Size: 8mm; Color: Yellow; Case of 1000
  • Extended controlled dropper tip dispenses one drop at a time
  • Average drop size is 44μL ±6μL
  • Case of 1000
Description & Specifications
Specifications
ISO 9001
41μL
1000 Cs.
Plastic dropping bottle
Yellow
Polyethylene
Dropper Tips