Please login to your online account to display your discounted pricing

Wheaton™ Low-Density Extended Controlled Dropper Tips (15mm)


Manufacturer: wheaton™  W242411A

 View more versions of this product

Catalog No. 03-440-57

  • / Case of 1000 Each
 

Description & Specifications

Specifications

For Use With (Equipment) Plastic dropping bottle
Color Pink
Certifications/Compliance ISO 9001
Material Polyethylene
No. per Case 1000 Cs.
Drop Size 41μL