missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Wheaton™ Leak-Resistant Wide-Mouth Natural PP Bottles

With double-seal closure with cap attached

Manufacturer:  Wheaton™ 209669

 View more versions of this product

Catalog No. 03-083-53


 

Specifications Description & Specifications

Specifications

16.9 oz., 500mL
16.9 oz.
Polypropylene
Double seal closure, no-drip pour lip
Polypropylene
53-410
72mm
Wide
Screw
500mL
ISO 9001
Natural
6.45 in.
Convenience
2.8 in.
164mm
Round