missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Wheaton™ Leak-Resistant Narrow-Mouth Natural PP Bottles

With double-seal closure with cap attached

Manufacturer:  Wheaton™ 209167

 View more versions of this product

Catalog No. 03-083-31



 

Specifications Description & Specifications

Specifications

4.22 oz., 125mL
4.22 oz.
Polypropylene
Double seal closure, no-drip pour lip
Polypropylene
24-410
50mm
Narrow
Screw
125mL
ISO 9001
Natural
3.97 in.
Convenience
1.96 in.
101mm
Round