missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Wheaton™ Leak-Resistant Amber HDPE Bottles with Caps
Ideal primary packaging solution for assays in diagnostic kits, and also chemical and storage purposes. With double-seal closure with cap attached.
$214.36 - $592.91
Specifications
Closure Material | Polypropylene |
---|---|
Closure Type | Screw Cap |
Color | Amber |
Material | HDPE |
Certifications/Compliance | ISO 9001 |
Catalog Number | Mfr. No. | Capacity | Height (Metric) | Neck Finish | Height (English) | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity | Height (Metric) | Neck Finish | Height (English) | Price | Quantity | ||
01183047
|
DWK Life Sciences
DWK209121 |
0.1 oz., 4mL | 38 mm | 13-425 | 1.4 in. |
Case of 72 Each for $256.44
|
|
||
03-083-18
|
DWK Life Sciences
209123 |
0.3 oz., 8 mL | 43 mm | 20-410 | 1.69 in. |
Case of 72 Each for $263.94
|
|
||
03-083-19
|
DWK Life Sciences
209124 |
0.5 oz., 15 mL | 56 mm | 20-410 | 2.2 in. |
Case of 72 Each for $214.36
|
|
||
03-083-20
|
DWK Life Sciences
209125 |
1 oz., 30mL | 69 mm | 20-410 | 2.71 in. |
Case of 72 Each for $309.55
|
|
||
03-083-21
|
DWK Life Sciences
209126 |
2 oz., 60 mL | 84 mm | 20-410 | 3.3 in. |
Case of 72 Each for $329.33
|
|
||
03-083-22
|
DWK Life Sciences
209127 |
4.2 oz., 125 mL | 101 mm | 24-410 | 3.9 in. |
Case of 72 Each for $417.18
|
|
||
03-083-23
|
DWK Life Sciences
209128 |
8.45 oz., 250 mL | 129 mm | 24-410 | 5.07 in. |
Case of 72 Each for $592.91
|
|
||
03-083-24
|
DWK Life Sciences
209129 |
17 oz., 500 mL | 171 mm | 28-410 | 6.73 in. |
Case of 48 Each for $582.21
|
|
||
03-083-25
|
DWK Life Sciences
209130 |
32 oz., 1000 mL | 210 mm | 38-430 | 8.2 in. |
Case of 24 Each for $476.79
|
|
||
Description
With double-seal closure with cap attached
Ideal for short- and long-term storage and packaging.
- Narrow-mouth
- No drip pour lip for clean pouring
- Polypropylene cap attached remains tight over an extended time
- Convenience packed
- Color: Amber
Specifications
Polypropylene | |
Amber | |
ISO 9001 |
Screw Cap | |
HDPE |