missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Wheaton™ Leak-Resistant Amber HDPE Bottles with Caps
Ideal primary packaging solution for assays in diagnostic kits, and also chemical and storage purposes. With double-seal closure with cap attached.
Supplier: DWK Life Sciences 209128
Description
With double-seal closure with cap attached
Ideal for short- and long-term storage and packaging.
- Narrow-mouth
- No drip pour lip for clean pouring
- Polypropylene cap attached remains tight over an extended time
- Convenience packed
Specifications
Polypropylene | |
61 mm | |
Screw Cap | |
8.45 oz., 250 mL | |
8.45 oz. | |
2.4 in. | |
24-410 |
129 mm | |
5.07 in. | |
24-410 mm | |
250 mL | |
Amber | |
HDPE | |
ISO 9001 |