Please login to your online account to display your discounted pricing

Wheaton™ Labeled Reagent Bottles


Manufacturer: wheaton™  220283

 View more versions of this product

Catalog No. 03-439-68B

  • / Case of 6 Each
 

Description & Specifications

Specifications

Capacity 16.9 oz., 500mL
Capacity (Metric) 500mL
Capacity (English) 16.9 oz.
Closure Material Polypropylene
Closure Liner Polyethylene
Color Clear
Diameter (English) 3.46 in.
Includes No-drip pour lip, large writing patch
Height (English) 7.48 in.
Material Glass
Diameter (Metric) 88mm
Height (Metric) 190mm
Mouth Wide
Closure Type Screw