Please login to your online account to display your discounted pricing
Wheaton™ Kuderna-Danish Evaporative Concentrator With Clear-Seal™ Joints
For concentration of trace amounts of analyte in organic solvent
$235.11 - $690.63
Specifications
Certifications/Compliance ASTM E 438
For Use With (Application) Concentration of trace amounts of analyte in organic solvent
View More Specs
Products
Catalog Number Mfr. No. Includes Type Price Quantity    
02-912-563
wheaton™
W297883
Clear seal joint Kuderna-Danish Evaporative Concentrator Case for $690.63
02-912-564
wheaton™
W384353
24/40 clear seal joint Three-ball Snyder Column Case for $322.89
02-912-565
wheaton™
W297825
24/40 top and 19/22 bottom clearseal joints Evaporative Flask Case for $235.11
Description & Specifications
Specifications
Certifications/Compliance ASTM E 438
For Use With (Application) Concentration of trace amounts of analyte in organic solvent
  • Smooth Clear-Seal joints eliminate need for grease, reducing possibility of contaminating sample
  • Complete unit consists of 270mm three-ball Snyder column, 500mL flask and 10mL receiving vessel
  • Manufactured from low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.