missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Wheaton™ Jars with Polypropylene Screw Cap

Manufacturer:  Wheaton™ WW209905

 View more versions of this product

Catalog No. 02-912-985


 

Specifications

Description

Jar; Wheaton; Durable; Chemical resistant; Pre-attached polypropylene screw cap unlined; With linerless autoclavable cap; Capacity: 1000mL; Color: white

  • Durable container with good chemical resistance
  • Pre-attached white polypropylene screw cap unlined or with foamed polyethylene liner
  • Polypropylene container with linerless cap is autoclavable
Description & Specifications

Specifications

32 oz., 1000mL
32 oz.
White
Jars
White
4.56 in.
97mm
1000mL
Polypropylene
3.81 in.
Yes
120-400
116mm
Screw