DWK Life Sciences Wheaton™ Impellers for Celstir™ Spinner Flasks
Manufacturer: DWK Life Sciences 356899
Description
- This non-breakable stainless steel shaft, for replacing the glass shaft included with Celstir™ flasks, may be retrofitted to the existing Celstir flasks.
Specifications
250mL Celstir™ Spinner Flask | |
Top cap with liners, glass rod, magnet holder, magnet and stirring paddle | |
Impeller |