DWK Life Sciences Wheaton™ Impellers for Celstir™ Spinner Flasks
Manufacturer:  DWK Life Sciences 356896
 View more versions of this product
Catalog No. 06-416-5C
Description
Description
  • This non-breakable stainless steel shaft, for replacing the glass shaft included with Celstir™ flasks, may be retrofitted to the existing Celstir flasks.
Specifications
Specifications
125mL Celstir™ Spinner Flask
Top cap with liners, glass rod, magnet holder, magnet and stirring paddle
Impeller