missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Wheaton™ Headspace Autosampler Vials: Screw Thread

Manufacturer:  Wheaton™ WW225286

 View more versions of this product

Catalog No. 03-410-729


 

Specifications Description & Specifications

Specifications

Headspace Autosampler Vial
20mL
Clear
75.5mm
23mm
Round
18mm
Borosilicate Glass
Screw cap
100 Case
2.97 in.
20mL