DWK Life Sciences Wheaton™ HDPE Serum Bottles
Manufacturer:  DWK Life Sciences 224031
 View more versions of this product
Catalog No. 06-446W
Description
Description
  • Manufactured from high density polyethylene
  • Widely used for animal health products and storage of non-medical products
  • Accepts standard rubber stoppers and aluminum seals
Specifications
Specifications
Rubber
2.2 in.
0.5 oz., 15 mL
0.5 oz.
0.78 in.
1.1 in.
13 mm
Serum Bottle
28 mm
56 mm
15 mL
Natural
0.51 in.
20 mm
HDPE