missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Wheaton™ Green Starline Polypropylene Caps for Redi-Pak™ Narrow-Mouth Containers
$58.83 - $110.26
Specifications
Certifications/Compliance | ISO 9001 |
---|---|
Color | Green |
Material | Polypropylene |
For Use With (Equipment) | Redi-pak™ leak resistant narrow mouth bottle |
Catalog Number | Mfr. No. | Neck Finish | No. per Case | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Neck Finish | No. per Case | Price | Quantity | ||||
09-877-11E
|
DWK Life Sciences
23950606 |
20-410 | 72 Cs. |
Case of 72 Each for $61.77
|
|
09-877-13E
|
DWK Life Sciences
23951206 |
28-410 | 48 Cs. |
Case of 48 Each for $110.26
|
|
09-877-10R
|
DWK Life Sciences
23950106 |
13-425 | 72 Cs. |
Case of 72 Each for $58.83
|
|
Description
- Leak resistant
- Autoclavable
Specifications
ISO 9001 | |
Green | |
Polypropylene | |
Redi-pak™ leak resistant narrow mouth bottle |