Please login to your online account to display your discounted pricing
Wheaton™ Graduated Valumetric™ Bottles: With White PP Cap With PTFE-Faced Foamed PE Liner
For storage and samples of solids, dry powdered materials and liquids
$138.51 - $240.37
Specifications
Closure Liner PTFE/Polyethylene
Closure Material Polypropylene
Closure Type Screw
Closure Color White
Color Clear
View More Specs
Products
Catalog Number Mfr. No. Capacity Closure Color Height (English) Height (Metric) Price Quantity    
02-912-218
wheaton™
W216972
2 oz., 60mL White 3.3 in. 85mm Case of 48 Each for $209.66
02-912-217
wheaton™
W216971
1 oz., 30mL White 2.7 in. 70mm Case of 48 Each for $185.80
02-912-220
wheaton™
W216974
8 oz., 250mL White 5 in. 129mm Case of 24 Each for $175.31
02-912-219
wheaton™
W216973
4 oz., 125mL White 4 in. 104mm Case of 24 Each for $138.51
02-912-221
wheaton™
W216975
16 oz., 500mL White 5.7 in. 147mm Case of 24 Each for $240.37
Description & Specifications
Specifications
Closure Liner PTFE/Polyethylene
Closure Material Polypropylene
Closure Type Screw
Closure Color White
Color Clear
  • Ideal for use as sample bottles
  • Useful for dry powders, soil samples, and specimen storage
  • Made from Wheaton 800 glass, which meets USP Type III standards
  • Graduated in mL and oz.
Autoclavable black phenolic cap with rubber liner.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.