Please login to your online account to display your discounted pricing
Wheaton™ Glass Staining Dish for 5 or 10 Slides
For fully immersed slides
$56.70 - $104.97
Specifications
Closure Type Screw
View More Specs
Includes:  Linerless polypropylene screw cap to reduce evaporation
Products
Catalog Number Mfr. No. Type Material Price Quantity    
08-813E
wheaton™
900570
Slide Staining Jar Soda Lime Glass Each for $56.70
Case of 6 Each for $340.20
02-912-618
wheaton™
W900522
Linerless Screw Cap Polypropylene Case of 6 Each for $104.97
Description & Specifications
Specifications
Closure Type Screw
  • Coplin staining jar holds up to five single slides or ten slides 3 × 1 in. (75 × 25mm) back-to-back vertically
  • 08-813E: Dish made from soda-lime glass
  • 02-912-618: Replacement cap: polypropylene
Outside height without cap: 4.25 in. (108mm). Slide grip available separately (08-813-1)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.