DWK Life Sciences Wheaton™ Glass Sample Vials in Lab File With Caps Attached
Attached caps help maintain cleanliness
Manufacturer:  DWK Life Sciences 224981
 View more versions of this product
Catalog No. 06-412-19
Description
Description
Attached caps help maintain cleanliness
Lab File with partitioned trays provides an easy way to store empty vials or to inventory samples.
  • Offer a variety of uses ranging from sample storage to freeze-drying
  • Fabricated from glass tubing for uniform sidewall and bottom thickness
  • Vials and caps are autoclavable
  • Amber vials made from borosilicate glass to protect light-sensitive products
Cap and Cap Liners
  • Solid top, black phenolic caps
  • 14B rubber-lined black phenolic screw cap for aqueous samples
  • PTFE/14B rubber-lined black phenolic screw cap for organic samples
Specifications
Specifications
Amber
Borosilicate Glass
Low profile for greater stability and less dead volume above the sample, making it ideal for compound storage
Screw
Phenolic
Screw Cap
USP Type I
12 mm
2 mL
14B Rubber
0.5 dr.
Autoclavable
38 mm
M-T File™ and Vial Rack