Please login to your online account to display your discounted pricing

Wheaton™ Glass Diagnostic Vials with Screw Threads


Alternative to serum bottles and vials

Manufacturer: wheaton™  WW219365

 View more versions of this product

Catalog No. 02-912-450

  • / Case of 480 Each
 

Description & Specifications

Specifications

Color Clear
Type Diagnostic Vials
Volume (Metric) 5mL
Grade Type 1
Neck Style Screw Thread
Volume (English) 0.17

Ideal for lyophilization.

  • Screw-thread design eliminates the need for crimping
  • Fits with I-Loc™ closure or screw cap with thin flange stopper
  • Tubular design provides excellent clarity and dimensional consistency from vial to vial
  • Heavy-walled glass provides impact resistance