DWK Life Sciences Wheaton™ Glass Diagnostic Vials with Screw Threads
Alternative to serum bottles and vials
Description
Ideal for lyophilization.
- Screw-thread design eliminates the need for crimping
- Fits with I-Loc™ closure or screw cap with thin flange stopper
- Tubular design provides excellent clarity and dimensional consistency from vial to vial
- Heavy-walled glass provides impact resistance
Specifications
Amber | |
Type 1 | |
0.68 |
20 mL | |
Screw | |
Diagnostic Vial |