Wheaton™ Glass Diagnostic Vials with Screw Threads
Alternative to serum bottles and vials
Manufacturer:  Wheaton™ WW219375
 View more versions of this product
Catalog No. 02-912-451
Specifications
Description
Vials, Diagnostic; Wheaton; Type 1 glass; Screw thread; Amber; 5mL
Ideal for lyophilization.
  • Screw-thread design eliminates the need for crimping
  • Fits with I-Loc™ closure or screw cap with thin flange stopper
  • Tubular design provides excellent clarity and dimensional consistency from vial to vial
  • Heavy-walled glass provides impact resistance
Description & Specifications
Specifications
Amber
5mL
Screw Thread
Diagnostic Vials
Type 1
0.17