missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Wheaton™ FMK Tubing

Manufacturer:  DWK Life Sciences WI054343

 View more versions of this product

Catalog No. 02-913-311



Description

Description

  • FKM is also a standout material in its resistance to weather-related aging and ozone
  • FKM tubing is sold in 1 foot (0.3 m) increments.
Specifications

Specifications

Black
3mm
0.169 in.
25 ft.
0.09 in.
Pumps and tubing
0.188 in.
FMK
4.3mm
7.62m
2.3mm
Documents