Wheaton™ Field Installed Rotation Alarm
Manufacturer:  Wheaton™ W349016
 View more versions of this product
Catalog No. 02-912-605
Specifications
Description
Alarm, Rotation; Wheaton; Field installation; Optional for cost buster production roller; Monitors drive system; Warning sound; Visual and audible; Remote signal: four pin panel mount mic jack; Pulsating, dry contact closure 5A at 24V DC
  • Protects cells during power outages
  • Provides a minimum of 24 hours of auxiliary power
  • Can be used to rotate vessels during transport to harvest area
Description & Specifications
Specifications
Audible and visual
Cost buster production roller culture
24VDC
5A
Field Installed Rotation Alarm