Please login to your online account to display your discounted pricing

Wheaton™ Extended Controlled 20mm LDPE Dropper Tips


Manufacturer: wheaton™  W242440A

 View more versions of this product

Catalog No. 03-440-25

  • / Case of 1000 Each
 

Description & Specifications

Specifications

Drop Size 41μL
Certifications/Compliance ISO 9001
Color White
For Use With Plastic dropping bottle
Material Polyethylene
No. per Case 1000 Cs.