Wheaton™ Extended Controlled 20mm LDPE Dropper Tips
Manufacturer:  Wheaton™ W242431A
 View more versions of this product
Catalog No. 03-440-35
Specifications
Description
Caps, Extended Controlled Dropper Tip; Wheaton; LDPE; Size: 20mm; Color: Pink; Case of 1000
  • Extended controlled dropper tip dispenses one drop at a time
  • Average drop size is 44μL ±6μL
  • Case of 100
Description & Specifications
Specifications
ISO 9001
41μL
1000 Cs.
Plastic dropping bottle
Pink
Polyethylene
Dropper Tips