Please login to your online account to display your discounted pricing

Wheaton™ Extended Controlled 20mm LDPE Dropper Tips


Manufacturer: wheaton™  W242433A

 View more versions of this product

Catalog No. 03-440-33

  • / Case of 1000 Each
 

Description & Specifications

Specifications

For Use With (Equipment) Plastic dropping bottle
Certifications/Compliance ISO 9001
Color Orange
Drop Size 41μL
Material Polyethylene
No. per Case 1000 Cs.