DWK Life Sciences Wheaton™ Extended Controlled 20mm LDPE Dropper Tips
Manufacturer:  DWK Life Sciences W242434A
 View more versions of this product
Catalog No. 03-440-32
Description
Description
  • Extended controlled dropper tip dispenses one drop at a time
  • Average drop size is 44μL ±6μL
  • Case of 100
Specifications
Specifications
ISO 9001
41μL
1000 Cs.
Plastic dropping bottle
Blue
Polyethylene
Dropper Tips
Documents